Trypan blue
Supplier: TCI America
T0556-25G
TCT0556-025GEA
39.69
USD
TCT0556-025G
Trypan blue
Trypan Blue
CAS Number: 72-57-1
MDL Number: MFCD00003969
Molecular Formula: C34H28N6O14S4
Molecular Weight: 960.79
Form: Crystal
Color: Deep Green
Other Product Names / Synonyms: Trypan Blue
Formula:
C₃₄H₂₄N₆Na₄O₁₄S₄ |
MDL Number:
MFCD00003969 CAS Number: 72-57-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...