DL-Tyrosine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 556-03-6
MDL Number: MFCD00063074
Molecular Formula: C9H11NO3
Molecular Weight: 181.19
Purity/Analysis Method:
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Tyr-OH
Formula:
C₉H₁₁NO₃ Boiling Pt: 378 °C (1013 hPa) Melting Pt: 285 °C |
MDL Number:
MFCD00063074 CAS Number: 556-03-6 EINECS: 209-113-1 Merck Index: 12,09970 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...