Melamine ≥98.0%

Supplier: TCI America

Synonyms: 2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine

T0337-25G T0337-500G
TCT0337-25GEA 16.79 USD
TCT0337-25G TCT0337-500G
Melamine ≥98.0%
Melamine

CAS Number: 108-78-1 MDL Number: MFCD00006055 Molecular Formula: C3H6N6 Molecular Weight: 126.12 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Flash Point (°C): 287


Other Product Names / Synonyms: 2,4,6-Triamino-1,3,5-triazine Monomer

Formula: C₃H₆N₆
MW: 126.12 g/mol
Density: 1.57 g/cm³ (20 °C)
Flash Pt: 287 °C
MDL Number: MFCD00006055
CAS Number: 108-78-1
EINECS: 203-615-4
Merck Index: 13,05834
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=096489abad01310cdf0251353fd7b3e9dc02d5ca&vid=50ed378d79a3049fc269e73a40809d4a2cf7e4bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR