p-Toluenesulfonic acid monohydrate ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate, 4-Methylbenzenesulfonic acid hydrate, Toluene-4-sulfonic acid monohydrate
CAS Number: 6192-52-5
MDL Number: MFCD00064387
Molecular Formula: C7H8O3S
Molecular Weight: 172.20
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Sulfonic Acids, Lewis Acids (Multi-component Reaction)
Other Product Names / Synonyms: PTSA
Formula:
C₇H₈O₃S·1H₂O Boiling Pt: 140 °C Melting Pt: 106 °C Density: 1.24 g/cm³ (20 °C) Flash Pt: 184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142137 CAS Number: 6192-52-5 EINECS: 203-180-0 UN: 2585 ADR: 8,III Merck Index: 12,09671 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...