Toluene ≥99.5%

Supplier: TCI America
Danger

T0260-100ML
TCT0260-100MLEA 62.35 USD
TCT0260-100ML
Toluene ≥99.5%
Toluene

[for Spectrophotometry] CAS Number: 108-88-3 MDL Number: MFCD00008512 Molecular Formula: C7H8 Molecular Weight: 92.14 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 110 Melting point (°C): -95 Flash Point (°C): 5 Specific Gravity (20/20): 0.87

Formula: C₆H₅CH₃
Boiling Pt: 110 °C
Melting Pt: –95 °C
Density: 0.87 (20 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR