Thiophene ≥98.0%

Supplier: TCI America
Danger

T0223-500ML T0223-25ML
TCT0223-500MLEA 72.78 USD
TCT0223-500ML TCT0223-25ML
Thiophene ≥98.0%
Thiophene

CAS Number: 110-02-1 MDL Number: MFCD00005413 Molecular Formula: C4H4S Molecular Weight: 84.14 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 84 Melting point (°C): -38 Flash Point (°C): -1 Specific Gravity (20/20): 1.07

Formula: C₄H₄S
Boiling Pt: 84 °C
Melting Pt: –38 °C
Density: 1.07 (20 °C)
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00005413
CAS Number: 110-02-1
EINECS: 203-729-4
UN: 2414
ADR: 3,II
Merck Index: 12,09490
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cf585f00416deda1c555da762e733ee399fe29a&vid=1cf585f00416deda1c555da762e733ee399fe29a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR