6-Thioguanine ≥95.0% (by HPLC)

Supplier: TCI America

T0212-1G T0212-5G
TCT0212-1GEA 95.65 USD
TCT0212-1G TCT0212-5G
6-Thioguanine ≥95.0% (by HPLC)
6-Thioguanine

CAS Number: 154-42-7 MDL Number: MFCD00233553 Molecular Formula: C5H5N5S Molecular Weight: 167.19 Purity/Analysis Method: 95.0% (HPLC) Form: Crystal Color: Slightly Pale Yellow


Product Categories: Bioscience, Immunosuppressant Ingredients, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleobases and Their Analogs Other Product Names / Synonyms: 2-Amino-6-mercaptopurine, 6-Mercaptoguanine

Formula: C₅H₅N₅S
MDL Number: MFCD00233553
CAS Number: 154-42-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca20e751f24bdf9906c361dcf6236b1ac2d953ae&vid=ca20e751f24bdf9906c361dcf6236b1ac2d953ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR