2,6,10,14-Tetramethylpentadecane ≥95.0%

Supplier: TCI America

Synonyms: Pristane

T0149-5ML
TCT0149-5MLEA 128.81 USD
TCT0149-5ML
2,6,10,14-Tetramethylpentadecane ≥95.0%
2,6,10,14-Tetramethylpentadecane

CAS Number: 1921-70-6 MDL Number: MFCD00008952 Molecular Formula: C19H40 Molecular Weight: 268.53 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 299 Melting point (°C): -99 Flash Point (°C): 110 Specific Gravity (20/20): 0.78


Product Categories: Bioscience, Reagents for Carcinogenic Models, Reagents for Inflammation & Immunity Disorder Models Other Product Names / Synonyms: Pristane

Formula: C₁₉H₄₀
MW: 268.53 g/mol
Boiling Pt: 296
Melting Pt: –99 °C
Density: 0.78 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008952
CAS Number: 1921-70-6
Merck Index: 12,07932
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4ccc8b6b3cd524c71d1a793cb5facfc210c7791&vid=f4ccc8b6b3cd524c71d1a793cb5facfc210c7791&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR