N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

T0147-500ML T0147-25ML T0147-100ML
TCT0147-500MLEA 77.33 USD
TCT0147-500ML TCT0147-25ML TCT0147-100ML
N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥98.0% (by GC, titration analysis)
N,N,N',N'-Tetramethylethylenediamine (TEMED)

CAS Number: 110-18-9 MDL Number: MFCD00008335 Molecular Formula: C6H16N2 Molecular Weight: 116.21 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Color: Colorless Boiling point (°C): 122 Melting point (°C): -55 Flash Point (°C): 17 Specific Gravity (20/20): 0.78


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Ligands, Ligands ([2+2+2]Cycloaddition Reaction) Other Product Names / Synonyms: TEMED, TMEDA

Formula: C₆H₁₆N₂
Boiling Pt: 122 °C
Melting Pt: –55 °C (lit.)
Density: 0.78 (20 °C)
Flash Pt: 17 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c0de745f7152d5ecbf9919ef6115ae409e66df4&vid=4c0de745f7152d5ecbf9919ef6115ae409e66df4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)
Specific Gravity (20/20) 0.7750 - 0.7780
Refractive Index n20/D 1.4160 to 1.4190


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR