Erythrosin B ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: Tetraiodofluorescein, Solvent Red 140, C.I. 45430:2

T0124-25G
TCT0124-025GEA 175.79 USD
TCT0124-025G
Erythrosin B ≥95.0% (by HPLC)
Erythrosin B

CAS Number: 15905-32-5 MDL Number: MFCD00005044 Molecular Formula: C20H8I4O5 Molecular Weight: 835.90 Purity/Analysis Method: 95.0% (HPLC) Form: Crystal Color: Yellow


Form: Crystal
Color: Yellow
Product Categories: Materials Chemistry, Xanthene Dyes (Functional Organic Dyes), Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Iodoeosine

Formula: C₂₀H₈I₄O₅
MW: 835.89 g/mol
MDL Number: MFCD00005044
CAS Number: 15905-32-5
EINECS: 240-046-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af30453a3162a340c12c7482c7eb2519f63df355&vid=af30453a3162a340c12c7482c7eb2519f63df355&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR