Sulfolane ≥99.0%
Supplier: TCI America
|
Warning
|
CAS Number: 126-33-0
MDL Number: MFCD00005484
Molecular Formula: C4H8O2S
Molecular Weight: 120.17
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Sulfolane, Tetramethylene Sulfone
Formula:
C₄H₈O₂S Boiling Pt: 285 °C Flash Pt: 168 °C |
MDL Number:
MFCD00005484 CAS Number: 126-33-0 EINECS: 204-783-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...