Tetrahydrofuran ≥99.5% stabilized

Supplier: TCI America
Danger


Stabilization: Stabilised with Stabilised

T0104-25ML T0104-500ML
TCT0104-25MLEA 18 USD
TCT0104-25ML TCT0104-500ML
Tetrahydrofuran ≥99.5% stabilized
Tetrahydrofuran

(stabilized with BHT)
CAS Number: 109-99-9
MDL Number: MFCD00005356
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 65
Melting point (°C): -108
Flash Point (°C): -15
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: THF

Formula: C₄H₈O
Boiling Pt: 65 °C
Melting Pt: –108 °C
Density: 0.89 (20 °C)
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=569692eb71092b087c49e3c28c49ec6418c6bc69&vid=569692eb71092b087c49e3c28c49ec6418c6bc69&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR