1,1,2,2-Tetrachloroethane ≥97.0%

Supplier: TCI America
Danger

T0063-500G T0063-25G
TCT0063-500GEA 48.1 USD
TCT0063-500G TCT0063-25G
1,1,2,2-Tetrachloroethane ≥97.0%
1,1,2,2-Tetrachloroethane

CAS Number: 79-34-5 MDL Number: MFCD00000848 Molecular Formula: C2H2Cl4 Molecular Weight: 167.84 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 146 Melting point (°C): -44 Specific Gravity (20/20): 1.60


Other Product Names / Synonyms: Acetylene Tetrachloride

Formula: Cl₂HCCHCl₂
Boiling Pt: 146 °C
Melting Pt: –44 °C
Density: 1.60 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000848
CAS Number: 79-34-5
EINECS: 201-197-8
UN: 1702
ADR: 6.1,II
Merck Index: 13,09264
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f145031abedf97ad5c66e1ad218cc0f7459f9b2d&vid=a8fd02f4f04dfb2e7f5c0352998482a7ad98f01b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR