Tetrabutylammonium iodide ≥98.0%

Supplier: TCI America
Warning

Synonyms: Tetra-n-butylammonium iodide, TBAI

T0057-500G T0057-25G T0057-100G
TCT0057-500GEA 301.46 USD
TCT0057-500G TCT0057-025G TCT0057-100G
Tetrabutylammonium iodide ≥98.0%
Tetrabutylammonium iodide

CAS Number: 311-28-4 MDL Number: MFCD00011636 Molecular Formula: C16H36IN Molecular Weight: 369.38 Purity/Analysis Method: 98.0% (T) Form: Crystal Color: White Melting point (°C): 147


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Materials Chemistry, Secondary Battery Research Reagents Other Product Names / Synonyms: TBAI

Formula: C₁₆H₃₆IN
Melting Pt: 147 °C
Density: 1.0027 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011636
CAS Number: 311-28-4
EINECS: 206-220-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff33db41995de2c3207c21775c1ca7e01d30b0&vid=5fff33db41995de2c3207c21775c1ca7e01d30b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR