Tetrabutylammonium iodide ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Tetra-n-butylammonium iodide, TBAI
T0057-500G
T0057-25G
T0057-100G
TCT0057-500GEA
301.46
USD
TCT0057-500G
TCT0057-025G
TCT0057-100G
Tetrabutylammonium iodide ≥98.0%
Tetrabutylammonium iodide
CAS Number: 311-28-4
MDL Number: MFCD00011636
Molecular Formula: C16H36IN
Molecular Weight: 369.38
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: TBAI
Formula:
C₁₆H₃₆IN Melting Pt: 147 °C Density: 1.0027 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011636 CAS Number: 311-28-4 EINECS: 206-220-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...