Solvent red 197 ≥98.0%
Supplier: TCI America
Synonyms:
3-(Diethylamino)-7-imino-7H-benzo[4,5]imidazo[1,2-a]chromeno[3,2-c]pyridine-6-carbonitrile
CAS Number: 52372-39-1
MDL Number: MFCD02176212
Molecular Formula: C23H19N5O
Molecular Weight: 381.44
Purity/Analysis Method:
Product Categories: Materials Chemistry, Molecular Conductors, Coumarin Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: 3-(Diethylamino)-7-imino-7H-benzo[4,5]imidazo[1,2-a]chromeno[3,2-c]pyridine-6-carbonitrile
Formula:
C₂₃H₁₉N₅O |
MDL Number:
MFCD02176212 CAS Number: 52372-39-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...