D-erythro-Sphingosine ≥96.0%
Supplier: TCI America
Synonyms:
D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol
CAS Number: 123-78-4
MDL Number: MFCD00036751
Molecular Formula: C18H37NO2
Molecular Weight: 299.50
Purity/Analysis Method:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: (2S,3R,E)-2-Amino-4-octadecene-1,3-diol
Formula:
C₁₈H₃₇NO₂ MW: 299.50 g/mol Melting Pt: 74 °C |
MDL Number:
MFCD00036751 CAS Number: 123-78-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...