N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate, 2,5-Dioxopyrrolidin-1-yl 4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexanecarboxylate, 4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-succinimidyl ester, 4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, SMCC, Succinimidyl-trans-4-(N-maleimidylmethyl)cyclohexane-1-carboxylate

S0853-100MG S0853-1G
TCS0853-100MGEA 96.26 USD
TCS0853-100MG TCS0853-1G
N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate ≥98.0% (by HPLC)
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

CAS Number: 64987-85-5 MDL Number: MFCD00009634 Molecular Formula: C16H18N2O6 Molecular Weight: 334.33 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Color: White Melting point (°C): 177 Storage Temperature: 0°C


Product Categories: Bioscience, Crosslinkers Other Product Names / Synonyms: 4-(N-Maleimidomethyl)cyclohexanecarboxylic Acid N-Succinimidyl Ester, SMCC

Formula: C₁₆H₁₈N₂O₆
MW: 334.33 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00009634
CAS Number: 64987-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a95b812b75a71aa0353d45bed45a640ffd24174a&vid=a95b812b75a71aa0353d45bed45a640ffd24174a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR