N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate, 2,5-Dioxopyrrolidin-1-yl 4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexanecarboxylate, 4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-succinimidyl ester, 4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, SMCC, Succinimidyl-trans-4-(N-maleimidylmethyl)cyclohexane-1-carboxylate
CAS Number: 64987-85-5
MDL Number: MFCD00009634
Molecular Formula: C16H18N2O6
Molecular Weight: 334.33
Purity/Analysis Method:
Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: 4-(N-Maleimidomethyl)cyclohexanecarboxylic Acid N-Succinimidyl Ester, SMCC
Formula:
C₁₆H₁₈N₂O₆ MW: 334.33 g/mol Melting Pt: 177 °C |
MDL Number:
MFCD00009634 CAS Number: 64987-85-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...