Sulfanilamide ≥99.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: 4-Aminobenzenesulfonamide, 4-Aminobenzenesulphonamide

S0833-5G
TCS0833-5GEA 36.54 USD
TCS0833-5G
Sulfanilamide ≥99.0% (by titrimetric analysis) for biochemical research
Sulphanilamide

[for Biochemical Research]
CAS Number: 63-74-1
MDL Number: MFCD00007939
Molecular Formula: C6H8N2O2S
Molecular Weight: 172.20
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 166
Lambda max.: 257 nm (H2O)


Product Categories: Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 4-Aminobenzenesulfonamide

Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: 401
Melting Pt: 166 °C
MDL Number: MFCD00007939
CAS Number: 63-74-1
Merck Index: 13,09010
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4a46e1b4a7c5af9fc46ff5093a7ab1621766873&vid=564a4c33e7da6793a3438fd012f9d8aa35678d23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR