N-Succinimidyl Acrylate ≥98.0%
Supplier: TCI America
CAS Number: 38862-24-7
MDL Number: MFCD00078261
Molecular Formula: C7H7NO4
Molecular Weight: 169.14
Purity/Analysis Method:
Product Categories: Materials Chemistry, Monomers, Bioscience, Crosslinkers
Other Product Names / Synonyms: Acrylic Acid N-Succinimidyl Ester, N-Acryloyloxysuccinimide
Formula:
C₇H₇NO₄ Melting Pt: 71 °C |
MDL Number:
MFCD00078261 CAS Number: 38862-24-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...