Sodium formate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Formic acid sodium salt

S0807-500G
TCS0807-500GEA 38.65 USD
TCS0807-500G
Sodium formate ≥97.0% (by titrimetric analysis)
Sodium formate

CAS Number: 141-53-7 MDL Number: MFCD00013101 Molecular Formula: CH2O2 Molecular Weight: 68.01 Purity/Analysis Method: 97.0% (T) Form: Crystal Color: White Melting point (°C): 253


Other Product Names / Synonyms: Formic Acid Sodium Salt

Formula: HCO₂Na
MW: 68.01 g/mol
Melting Pt: 253 °C
Density: 1.919 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 13,08694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14463cb3654d78b8d6c10158c0c69a5d829e80c7&vid=14463cb3654d78b8d6c10158c0c69a5d829e80c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR