Silver sulfadiazine ≥98.0%
Supplier: TCI America
Synonyms:
sulfadiazine Silver, Sulphadiazine Silver
S0595-25G
S0595-5G
TCS0595-25GEA
188
USD
TCS0595-25G
TCS0595-5G
Silver sulfadiazine ≥98.0%
Silver sulfadiazine
CAS Number: 22199-08-2
MDL Number: MFCD00072101
Molecular Formula: C10H9AgN4O2S
Molecular Weight: 357.14
Purity/Analysis Method:
Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: Sulfadiazine Silver
Formula:
C₁₀H₉AgN₄O₂S Melting Pt: 285 °C |
MDL Number:
MFCD00072101 CAS Number: 22199-08-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...