Sodium benzoate ≥98.0%

Supplier: TCI America

Synonyms: Benzoic acid sodium salt

S0593-500G S0593-25G
TCS0593-500GEA 38 USD
TCS0593-500G TCS0593-25G
Sodium benzoate ≥98.0%
Sodium benzoate

CAS Number: 532-32-1 MDL Number: MFCD00012463 Molecular Formula: C7H6O2 Molecular Weight: 144.10 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Melting point (°C): 450 Flash Point (°C): 100


Other Product Names / Synonyms: Benzoic Acid Sodium Salt

Formula: C₆H₅COONa
Melting Pt: 450 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ccdbf705cece0a6111fa02324b674e21f7723a9&vid=a7169716b3ad86c378069f610fc8d1a6d4f2ed63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR