n-Hexadecane

Supplier: TCI America
Danger

Synonyms: Hexadecane, Cetane

S0555-250ML
TCS0555-250MLEA 203.51 USD
TCS0555-250ML
n-Hexadecane
n-Hexadecane

Reference Material for Flash Point Certified by The Japan Petroleum Institute CAS Number: 544-76-3 Molecular Formula: C16H34 Molecular Weight: 226.45 Form: Clear Liquid Color: Colorless


Other Product Names / Synonyms: Hexadecane

Formula: H₃C(CH₂)₁₄CH₃
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.77331 g/cm³ (20 °C)
Flash Pt: 135 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6400277e4e705803f2e949050d7fd98f53088e2&vid=7965ea93db730e2357c64efe985988dbc2f3fe3f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR