n-Decane
Supplier: TCI America
Reference Material for Flash Point Certified by The Japan Petroleum Institute
CAS Number: 124-18-5
MDL Number: MFCD00008954
Molecular Formula: C10H22
Molecular Weight: 142.29
Form: Clear Liquid
Color: Colorless
Other Product Names / Synonyms: Decane
Formula:
H₃C(CH₂)₈CH₃ |
MDL Number:
MFCD00008954 CAS Number: 124-18-5 UN: 2247 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...