Sodium ethoxide ∼20% in ethanol

Supplier: TCI America

S0548-500ML
TCS0548-500MLEA 59.32 USD
TCS0548-500ML
Sodium ethoxide ∼20% in ethanol
Sodium ethoxide

CAS Number: 141-52-6
MDL Number: MFCD00012417
Molecular Formula: C2H5NaO
Molecular Weight: 68.05
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 9
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Sodium Ethylate

Formula: C₂H₅NaO
Density: 0.88 (20 °C)
Flash Pt: 9 °C
MDL Number: MFCD00012417
CAS Number: 141-52-6
UN: 3206
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3b8cb0f392fb6c7fc056a8eb05b830e6f826ef&vid=e2095150eb32406f718531502bebd45aefe34e6a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR