Sodium ethoxide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

S0547-100G
TCS0547-100GEA 47.89 USD
TCS0547-100G
Sodium ethoxide ≥95.0% (by titrimetric analysis)
Sodium ethoxide

CAS Number: 141-52-6 MDL Number: MFCD00012417 Molecular Formula: C2H5NaO Molecular Weight: 68.05 Purity/Analysis Method: 95.0% (T) Form: Crystal Color: White Melting point (°C): 260 Flash Point (°C): 30


Other Product Names / Synonyms: Sodium Ethylate

Formula: C₂H₅NaO
Melting Pt: 260 °C
Flash Pt: 30 °C
MDL Number: MFCD00012417
CAS Number: 141-52-6
UN: 3206
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ca7fe58fa4e802e01d8ccd60c502585d3d1f162&vid=8ca7fe58fa4e802e01d8ccd60c502585d3d1f162&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR