Sodium hydroxide 2 mol/L in water

Supplier: TCI America

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

S0543-500ML
TCS0543-500MLEA 18.32 USD
TCS0543-500ML
Sodium hydroxide 2 mol/L in water
Sodium hydroxide

CAS Number: 1310-73-2
MDL Number: MFCD00003548
Molecular Formula: HNaO
Molecular Weight: 40.00
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.09

Formula: NaOH
Density: 1.09 (20 °C)
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df31f52d5e0152665ef678ca97e6af3a25a53c97&vid=acbffc6c68c2968152070ecef8b46e17162cda7c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR