D-Biotin-N-succinimidyl ester ≥97.0%

Supplier: TCI America

Synonyms: Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide

S0491-100MG S0491-1G
TCS0491-100MGEA 120 USD
TCS0491-100MG TCS0491-1G
D-Biotin-N-succinimidyl ester ≥97.0%
D-Biotin-N-succinimidyl ester

CAS Number: 35013-72-0 MDL Number: MFCD00078531 Molecular Formula: C14H19N3O5S Molecular Weight: 341.38 Purity/Analysis Method: 97.0% (HPLC,N) Form: Crystal Color: White Melting point (°C): 140 Storage Temperature: 0°C


Product Categories: Bioscience, Biotins & Streptavidins Other Product Names / Synonyms: D-Biotin N-Succinimidyl Ester, N-(D-Biotinyloxy)succinimide

Formula: C₁₄H₁₉N₃O₅S
Melting Pt: 140 °C
MDL Number: MFCD00078531
CAS Number: 35013-72-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ccde0dab244f5d9817f201bd09425300361cc60&vid=3ccde0dab244f5d9817f201bd09425300361cc60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR