Silver tetrafluoroborate ≥98.0%

Supplier: TCI America

Synonyms: Argentous tetrafluoroborate

S0446-25G S0446-5G
TCS0446-25GEA 298 USD
TCS0446-25G TCS0446-5G
Silver tetrafluoroborate ≥98.0%
Silver tetrafluoroborate

CAS Number: 14104-20-2 MDL Number: MFCD00003408 Molecular Formula: AgBF4 Molecular Weight: 194.67 Purity/Analysis Method: 98.0% (T) Form: Crystal Color: White Melting point (°C): 73

Formula: AgBF₄
Melting Pt: 73 °C
MDL Number: MFCD00003408
CAS Number: 14104-20-2
UN: 3260
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fe4a65f62d550db9aa823db1a776fbdd8be5a55&vid=8fe4a65f62d550db9aa823db1a776fbdd8be5a55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR