Sodium triacetoxyborohydride ≥80.0%
Supplier: TCI America
S0394-100G
S0394-25G
TCS0394-100GEA
203.99
USD
TCS0394-100G
TCS0394-25G
Sodium triacetoxyborohydride ≥80.0%
Sodium triacetoxyborohydride
CAS Number: 56553-60-7
MDL Number: MFCD00012211
Molecular Formula: C6H11BO6
Molecular Weight: 211.94
Purity/Analysis Method:
Formula:
C₆H₁₀BNaO₆ Melting Pt: 106 °C |
MDL Number:
MFCD00012211 CAS Number: 56553-60-7 UN: 3396 ADR: 4.3,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...