Sodium hexafluorophosphate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

S0392-25G
TCS0392-25GEA 121.09 USD
TCS0392-25G
Sodium hexafluorophosphate ≥98.0% (by titrimetric analysis)
Sodium hexafluorophosphate

CAS Number: 21324-39-0 MDL Number: MFCD00011122 Molecular Formula: F6NaP Molecular Weight: 167.95 Purity/Analysis Method: 98.0% (T) Form: Crystal Color: White

Formula: F₆NaP
MW: 167.95 g/mol
MDL Number: MFCD00011122
CAS Number: 21324-39-0
UN: 3260
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b04d09b4087b98ecac9337c4869a42087d74d9c&vid=1b04d09b4087b98ecac9337c4869a42087d74d9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR