Methyl oleate ≥99.0%

Supplier: TCI America

Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

S0326-5ML
TCS0326-005MLEA 114.91 USD
TCS0326-005ML
Methyl oleate ≥99.0%
Methyl oleate

[Standard Material for GC] CAS Number: 112-62-9 MDL Number: MFCD00009578 Molecular Formula: C19H36O2 Molecular Weight: 296.50 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 161 Melting point (°C): -20 Flash Point (°C): 159 Storage Temperature: 0-10°C


Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols, Analytical Science, Fatty Acid Methyl Esters (GC Standard) Other Product Names / Synonyms: Oleic Acid Methyl Ester

Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: 161 °C
Melting Pt: –20 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR