Methyl stearate ≥99.5%

Supplier: TCI America

S0312-5ML
TCS0312-005MLEA 171.47 USD
TCS0312-005ML
Methyl stearate ≥99.5%
Methyl stearate

[Standard Material for GC] CAS Number: 112-61-8 MDL Number: MFCD00009005 Molecular Formula: C19H38O2 Molecular Weight: 298.51 Purity/Analysis Method: 99.5% (GC) Form: Crystal Color: White Flash Point (°C): 153 Freezing point (°C): 37


Product Categories: Analytical Science, Fatty Acid Methyl Esters (GC Standard) Other Product Names / Synonyms: Stearic Acid Methyl Ester

Formula: CH₃(CH₂)₁₆CO₂CH₃
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.007 g/cm³ (20 °C)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00009005
CAS Number: 112-61-8
EINECS: 203-990-4
Merck Index: 12,08959

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR