Methyl laurate ≥99.5%

Supplier: TCI America

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

S0309-5ML
TCS0309-005MLEA 60.7 USD
TCS0309-005ML
Methyl laurate ≥99.5%
Methyl laurate

[Standard Material for GC] CAS Number: 111-82-0 MDL Number: MFCD00008966 Molecular Formula: C13H26O2 Molecular Weight: 214.35 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 262 Melting point (°C): 5 Flash Point (°C): 134 Specific Gravity (20/20): 0.87


Product Categories: Analytical Science, Fatty Acid Methyl Esters (GC Standard) Other Product Names / Synonyms: Lauric Acid Methyl Ester

Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 266
Melting Pt: 4.5
Density: 0.87 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00008966
CAS Number: 111-82-0
EINECS: 203-911-3

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR