Methyl hexanoate ≥99.5%

Supplier: TCI America

Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester

S0304-5ML
TCS0304-005MLEA 111.9 USD
TCS0304-005ML
Methyl hexanoate ≥99.5%
Methyl hexanoate

[Standard Material for GC] CAS Number: 106-70-7 MDL Number: MFCD00009510 Molecular Formula: C7H14O2 Molecular Weight: 130.19 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 150 Flash Point (°C): 46 Specific Gravity (20/20): 0.88


Product Categories: Analytical Science, Fatty Acid Methyl Esters (GC Standard) Other Product Names / Synonyms: Hexanoic Acid Methyl Ester

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 151
Melting Pt: -63
Density: 0.88 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00009510
CAS Number: 106-70-7
EINECS: 203-425-1
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR