n-Pentacosane ≥99.0%

Supplier: TCI America

S0297-1G
TCS0297-001GEA 125.36 USD
TCS0297-001G
n-Pentacosane ≥99.0%
n-Pentacosane

[Standard Material for GC] CAS Number: 629-99-2 MDL Number: MFCD00009353 Molecular Formula: C25H52 Molecular Weight: 352.69 Purity/Analysis Method: 99.0% (GC) Form: Crystal Color: White Boiling point (°C): 254 Melting point (°C): 54 Flash Point (°C): 120

Formula: H₃C(CH₂)₂₃CH₃
Boiling Pt: 254 °C
Melting Pt: 54 °C
Density: 0.8012 g/cm³ (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00009353
CAS Number: 629-99-2
EINECS: 211-123-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f8fe722e0fb459e1d53a6640769e86a6738eb48&vid=149952a214a73633cc9c6801929c27e7ae8dcea4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR