n-Tetracosane ≥99.0%

Supplier: TCI America

Synonyms: N-tetracosane

S0296-1G
TCS0296-001GEA 36.93 USD
TCS0296-001G
n-Tetracosane ≥99.0%
n-Tetracosane

[Standard Material for GC] CAS Number: 646-31-1 MDL Number: MFCD00009352 Molecular Formula: C24H50 Molecular Weight: 338.66 Purity/Analysis Method: 99.0% (GC) Form: Crystal Color: White Boiling point (°C): 250 Melting point (°C): 51 Flash Point (°C): 194

Formula: H₃C(CH₂)₂₂CH₃
MW: 338.66 g/mol
Boiling Pt: 391.3
Melting Pt: 51 °C
Density: 0.7787 g/cm³ (20 °C)
Flash Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00009352
CAS Number: 646-31-1
EINECS: 211-474-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38796145f0fb30e7460737f289a965e6053f9721&vid=c3c92018359e7d1bb4ec449b672a9bf78573c3d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR