n-Hexadecane ≥99.5%

Supplier: TCI America
Danger

Synonyms: Hexadecane, Cetane

S0288-5ML
TCS0288-005MLEA 50 USD
TCS0288-005ML
n-Hexadecane ≥99.5%
n-Hexadecane

[Standard Material for GC] CAS Number: 544-76-3 MDL Number: MFCD00008998 Molecular Formula: C16H34 Molecular Weight: 226.45 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 287 Flash Point (°C): 136 Freezing point (°C): 17 Specific Gravity (20/20): 0.77

Formula: H₃C(CH₂)₁₄CH₃
Boiling Pt: 287 °C
Melting Pt: 18 °C
Density: 0.77 (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR