n-Pentadecane ≥99.5%

Supplier: TCI America
Danger

Synonyms: N-Pentadecane, Pentadecane

S0287-5ML
TCS0287-005MLEA 75.3 USD
TCS0287-005ML
n-Pentadecane ≥99.5%
n-Pentadecane

[Standard Material for GC] CAS Number: 629-62-9 MDL Number: MFCD00008990 Molecular Formula: C15H32 Molecular Weight: 212.42 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 271 Flash Point (°C): 120 Freezing point (°C): 10 Specific Gravity (20/20): 0.77

Formula: H₃C(CH₂)₁₃CH₃
Boiling Pt: 271 °C
Melting Pt: 13…15 °C
Density: 0.77 (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00008990
CAS Number: 629-62-9
EINECS: 211-098-1
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR