n-Tetradecane ≥99.5%

Supplier: TCI America
Danger

S0286-5ML
TCS0286-005MLEA 53.4 USD
TCS0286-005ML
n-Tetradecane ≥99.5%
n-Tetradecane

[Standard Material for GC] CAS Number: 629-59-4 MDL Number: MFCD00008986 Molecular Formula: C14H30 Molecular Weight: 198.39 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 253 Melting point (°C): 6 Flash Point (°C): 100 Specific Gravity (20/20): 0.76

Formula: H₃C(CH₂)₁₂CH₃
Boiling Pt: 253 °C
Melting Pt: 6 °C
Density: 0.76 (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS Number: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR