n-Decane ≥99.5% (by GC)

Supplier: TCI America

S0282-5ML
TCS0282-5MLEA 75.39 USD
TCS0282-5ML
n-Decane ≥99.5% (by GC)
n-Decane

[Standard Material for GC] CAS Number: 124-18-5 MDL Number: MFCD00008954 Molecular Formula: C10H22 Molecular Weight: 142.29 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 174 Flash Point (°C): 50 Specific Gravity (20/20): 0.73

Formula: H₃C(CH₂)₈CH₃
Boiling Pt: 174 °C
Density: 0.73 (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00008954
CAS Number: 124-18-5
UN: 2247
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=030c835c07071ed9a1f68366b1a728d7f42282bd&vid=b6989c9909c75b60e7c46c9af6579482d41927d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR