n-Octane ≥99.5% (by GC)

Supplier: TCI America

S0280-5ML
TCS0280-5MLEA 92.6 USD
TCS0280-5ML
n-Octane ≥99.5% (by GC)
n-Octane

[Standard Material for GC] CAS Number: 111-65-9 MDL Number: MFCD00009556 Molecular Formula: C8H18 Molecular Weight: 114.23 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 125 Melting point (°C): -57 Flash Point (°C): 14 Specific Gravity (20/20): 0.70

Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 125 °C
Melting Pt: –57 °C
Density: 0.70 (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00009556
CAS Number: 111-65-9
UN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7288145963a633cba7e7d5f240313511e6f0e22&vid=c96becbf94b2e070e7bb149653171d6313ab9fe4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR