n-Hexane ≥99.5% (by GC)

Supplier: TCI America

S0278-5ML
TCS0278-5MLEA 86 USD
TCS0278-5ML
n-Hexane ≥99.5% (by GC)
n-Hexane

[Standard Material for GC]
CAS Number: 110-54-3
MDL Number: MFCD00009520
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 68
Melting point (°C): -95
Flash Point (°C): -22
Specific Gravity (20/20): 0.66

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 68 °C
Melting Pt: –95 °C
Density: 0.66 (20 °C)
Flash Pt: –22 °C
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=447bda59a149be62858a5ab0bdf0c5ec3428a1d1&vid=1508c274d910dc122c9a5db07236717ab121dc73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR