Sodium Copper Chlorophyllin
Supplier: TCI America
S0271-25G
TCS0271-025GEA
83
USD
TCS0271-025G
Sodium Copper Chlorophyllin
Sodium copper chlorophyllin
CAS Number: 28302-36-5
MDL Number: MFCD00012149
Form: Crystal
Color: Deep Green
Product Categories: Bioscience, Antioxidants, Porphyrins
MDL Number:
MFCD00012149 CAS Number: 28302-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...