Sinigrin hydrate ≥98.0% (by HPLC)
Supplier: TCI America
CAS Number: 3952-98-5
MDL Number: MFCD00006616
Molecular Formula: C10H17NO9S2
Molecular Weight: 397.45
Purity/Analysis Method:
Formula:
C₁₀H₁₇NO₉S₂ Melting Pt: 127 °C |
MDL Number:
MFCD00006616 CAS Number: 3952-98-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...