Safranine O

Supplier: TCI America

S0145-25G S0145-500G
TCS0145-025GEA 38.67 USD
TCS0145-025G TCS0145-500G
Safranine O
Safranine O

CAS Number: 477-73-6 MDL Number: MFCD00011759 Form: Crystal Color: Yellow Red


Other Product Names / Synonyms: Safranine T, Safranine O

Formula: C₂₀H₁₉ClN₄
Density: 1.447 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR