Sulfanilamide ≥99.0%
Supplier: TCI America
Synonyms:
4-Aminobenzenesulfonamide, 4-Aminobenzenesulphonamide
S0119-500G
S0119-25G
S0119-100G
TCS0119-500GEA
196.92
USD
TCS0119-500G
TCS0119-025G
TCS0119-100G
Sulfanilamide ≥99.0%
Sulphanilamide
CAS Number: 63-74-1
MDL Number: MFCD00007939
Molecular Formula: C6H8N2O2S
Molecular Weight: 172.20
Purity/Analysis Method:
Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 4-Aminobenzenesulfonamide
Formula:
H₂NC₆H₄SO₂NH₂ MW: 172.21 g/mol Boiling Pt: 401 Melting Pt: 166 °C Density: 1.46 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007939 CAS Number: 63-74-1 EINECS: 200-563-4 Merck Index: 13,09010 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...