Butyl stearate ≥97.0%

Supplier: TCI America
Warning

Synonyms: Stearic acid butyl ester, Butyl octadecanoate

S0077-25G
TCS0077-025GEA 30.79 USD
TCS0077-025G TCS0077-400G
Butyl stearate ≥97.0%
Butyl stearate

CAS Number: 123-95-5 MDL Number: MFCD00026669 Molecular Formula: C22H44O2 Molecular Weight: 340.59 Purity/Analysis Method: 97.0% (GC) Form: Crystal Color: White Flash Point (°C): 160 Freezing point (°C): 25


Other Product Names / Synonyms: Stearic Acid Butyl Ester

Formula: C₂₂H₄₄O₂
MW: 340.59 g/mol
Boiling Pt: 220
Melting Pt: 21
Density: 0.854 g/cm³ (20 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00026669
CAS Number: 123-95-5
EINECS: 204-666-5
Merck Index: 13,01589
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fe0cadc953eb318802238677b378606c1e5941a&vid=0fe0cadc953eb318802238677b378606c1e5941a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR